Dimethyl itaconate ≥97%

Supplier: Thermo Fisher Scientific

L12652.22 L12652.36
L12652.22EA 52.9 EUR
L12652.22 L12652.36
Dimethyl itaconate ≥97%
Dimethyl itaconate

Purity: 97% CAS No.: 617-52-7 Molecular Formula: C7H10O4 Formula Weight: 158.16 Flash Point: 100°(212°F) Melting Point: 33-39° Boiling Point: 207-208° Beilstein Reference: 386674 MDL No.: MFCD00008446

Formula: C₇H₁₀O₄
Boiling Pt: 207…208 °C
Melting Pt: 33…39 °C
Density: 1,124
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008446
CAS Number: 617-52-7
EINECS: 210-519-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR