2,6-Dichlorophenyl isothiocyanate ≥98%

Supplier: Thermo Fisher Scientific
Danger

L12672.06 L12672.03
L12672.06EA 232 EUR
L12672.06 L12672.03
2,6-Dichlorophenyl isothiocyanate ≥98%
2,6-Dichlorophenyl isothiocyanate

Purity: 98+% CAS No.: 6590-95-0 Molecular Formula: C7H3Cl2NS Formula Weight: 204.08 Storage Temperature: 2°C to 8°C Odour: Irritating Boiling Point: 122-124°/8mm Beilstein Reference: 639061 UN No.: 2923 MDL No.: MFCD00041056

Formula: C₇H₃Cl₂NS
Boiling Pt: 122…124 °C (8 mmHg)
Melting Pt: 42…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041056
CAS Number: 6590-95-0
UN: 2923
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR