Diacetin mixture of isomers ∼50% remainder triacetin and monoacetin, tech.

Supplier: Thermo Fisher Scientific

L12708.30 L12708.0B
L12708.30EA 42.7 EUR
L12708.30 L12708.0B
Diacetin mixture of isomers ∼50% remainder triacetin and monoacetin, tech.
Diacetin

CAS No.: 25395-31-7 Molecular Formula: C7H12O5 Formula Weight: 176.17 Flash Point: 141°(285°F) Refractive Index: 1.4440 Boiling Point: 250-280° dec. Beilstein Reference: 1706903 MDL No.: MFCD00008717

Formula: C₇H₁₂O₅
Boiling Pt: 250…280 °C (dec.)
Density: 1,190
Flash Pt: 141 °C (285 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008717
CAS Number: 25395-31-7
EINECS: 246-941-2
Merck Index: 14,2961

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR