1-(Triisopropylsilyl)pyrrole 95%

Supplier: Thermo Fisher Scientific
Warning

L13152.03 L13152.06 L13152.14
L13152.03EA 0 EUR
L13152.03 L13152.06 L13152.14
1-(Triisopropylsilyl)pyrrole 95%
1-(Triisopropylsilyl)pyrrole

Purity: 95% CAS No.: 87630-35-1 Molecular Formula: C13H25NSi Formula Weight: 223.44 Flash Point: 106°(222°F) Refractive Index: 1.4920 Boiling Point: 78°/0.4mm Beilstein Reference: 3540663 MDL No.: MFCD00054932

Formula: C₁₃H₂₅NSi
Boiling Pt: 78 °C (0,4 mmHg)
Density: 0,904
Flash Pt: 106 °C (222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054932
CAS Number: 87630-35-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR