Borane-tert-butylamine complex 97%

Supplier: Thermo Fisher Scientific
Danger

L13177.09 L13177.18
L13177.09EA 41.7 EUR
L13177.09 L13177.18
Borane-tert-butylamine complex 97%
Borane tert-butylamine complex

Purity: 97% CAS No.: 7337-45-3 Molecular Formula: (CH3)3CNH2.BH3 Formula Weight: 86.98 Melting Point: 96-98° Beilstein Reference: 4134905 UN No.: 2811 MDL No.: MFCD00075635

Formula: C₄H₁₄BN
Melting Pt: 96…98 °C
Density: 0,870
Storage Temperature: Ambient
MDL Number: MFCD00075635
CAS Number: 7337-45-3
EINECS: 230-851-5
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR