2-(2-Ethoxyethoxy)ethyl acetate 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diethylene glycol monoethyl ether acetate

L13446.AE L13446.AP L13446.0F
L13446.AEEA 28 EUR
L13446.AE L13446.AP L13446.0F
2-(2-Ethoxyethoxy)ethyl acetate 99
2-(2-Ethoxyethoxy)ethyl acetate

Purity: 99% CAS No.: 112-15-2 Molecular Formula: CH3CO2(CH2CH2O)2C2H5 Formula Weight: 176.21 Flash Point: 95°(203°F) Refractive Index: 1.4210 Melting Point: -25° Boiling Point: 218-219° Beilstein Reference: 1764643 Merck Reference: 14,1800 MDL No.: MFCD00041928

Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 218…219 °C
Melting Pt: –25 °C
Density: 1.012
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041928
CAS Number: 112-15-2
EINECS: 203-940-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR