Dowex® 50WX8 (H) 200-400 mesh

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Dowex 50 wx8

L13922.18 L13922.30 L13922.0B
L13922.18EA 70.6 EUR
L13922.18 L13922.30 L13922.0B
Dowex® 50WX8 (H) 200-400 mesh
Dowex® 50WX8 (H)

CAS No.: 11119-67-8 MDL No.: MFCD00132726

Storage Temperature: Ambient
MDL Number: MFCD00132726
CAS Number: 11119-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR