Silica gel, Pore size: 60 Å 0.032-0.063 mm (230-450 mesh)

Supplier: Thermo Fisher Scientific
Warning

L14003.36 L14003.0B L14003.0E
L14003.36EA 70.2 EUR
L14003.36 L14003.0B L14003.0E
Silica gel, Pore size: 60 Å 0.032-0.063 mm (230-450 mesh)
Silica gel

CAS No.: 7631-86-9 Odour: Odourless Merck Reference: 14,8493 MDL No.: MFCD00011232

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR