N-(3-(Trimethoxysilyl)propyl)ethylenediamine 96%

Supplier: Thermo Fisher Scientific
Danger

L14043.14 L14043.22 L14043.36
L14043.14EA 35.8 EUR
L14043.14 L14043.22 L14043.36
N-(3-(Trimethoxysilyl)propyl)ethylenediamine 96%
N-(3-(Trimethoxysilyl)propyl)ethylenediamine

Purity: 96% CAS No.: 1760-24-3 Molecular Formula: (CH3O)3Si(CH2)3NHCH2CH2NH2 Formula Weight: 222.36 Flash Point: 136°(276°F) Refractive Index: 1.4435 Odour: Ammonia-like Boiling Point: 261-263° Beilstein Reference: 636230 UN No.: 2735 MDL No.: MFCD00008173

Formula: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
Boiling Pt: 261…263 °C
Melting Pt: ∼ 0 °C
Density: 1,010
Flash Pt: 136 °C (276 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008173
CAS Number: 1760-24-3
EINECS: 217-164-6
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR