4-(Trifluoromethyl)benzhydrol ≥97%

Supplier: Thermo Fisher Scientific
Warning

L14147.03 L14147.06
L14147.03EA 18.6 EUR
L14147.03 L14147.06
4-(Trifluoromethyl)benzhydrol ≥97%
4-(Trifluoromethyl)benzhydrol

Purity: 97% CAS No.: 395-23-3 Molecular Formula: C14H11F3O Formula Weight: 252.23 Melting Point: 59-61° Boiling Point: 122-124°/0.4mm MDL No.: MFCD00014399

Formula: C₁₄H₁₁F₃O
Boiling Pt: 122…124 °C (0,4 mmHg)
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00014399
CAS Number: 395-23-3
EINECS: 209-917-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR