9-Bromo-9-phenylfluorene 96%

Supplier: Thermo Fisher Scientific
Danger

L14225.03 L14225.06
L14225.03EA 42.6 EUR
L14225.03 L14225.06
9-Bromo-9-phenylfluorene 96%
9-Bromo-9-phenylfluorene

Purity: 96% CAS No.: 55135-66-5 Molecular Formula: C19H13Br Formula Weight: 321.23 Melting Point: 94-99° Beilstein Reference: 2561445 UN No.: 3261 MDL No.: MFCD00075522

Formula: C₁₉H₁₃Br
Melting Pt: 94…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00075522
CAS Number: 55135-66-5
UN: 3261
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR