(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (+)-Mosher's Acid Chloride, (+)-MTPA-Cl, (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, (S)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoyl chloride

L14332.MF L14332.MD L14332.03
L14332.MFEA 47.7 EUR
L14332.MF L14332.MD L14332.03
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

Purity: 98+% CAS No.: 20445-33-4 Molecular Formula: C10H8ClF3O2 Formula Weight: 252.62 Storage Temperature: -30°C to -10°C Flash Point: 89°(192°F) Refractive Index: 1.4690 Boiling Point: 213-214° Beilstein Reference: 3591564 Optical Rotation: +137° (c=6.4 in carbon tetrachloride) UN No.: UN3265 MDL No.: MFCD00067105

Formula: C₁₀H₈ClF₃O₂
MW: 252.62 g/mol
Boiling Pt: 213…214 °C
Density: 1.353
Flash Pt: 89 °C (192 °F)
Storage Temperature: Freezer
MDL Number: MFCD00067105
CAS Number: 20445-33-4
UN: 3265
ADR: 8 II
Merck Index: 14,6280

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR