Methyl indole-3-carboxylate ≥99%

Supplier: Thermo Fisher Scientific

L14516.14 L14516.22
L14516.14EA 53 EUR
L14516.14 L14516.22
Methyl indole-3-carboxylate ≥99%
Methyl indole-3-carboxylate

Purity: 99% CAS No.: 942-24-5 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 149-152° Beilstein Reference: 142023

Formula: C₁₀H₉NO₂
Melting Pt: 149…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00189407
CAS Number: 942-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR