(3-Iodophenyl)acetic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

L15117.04 L15117.ME
L15117.04EA 136 EUR
L15117.04 L15117.ME
(3-Iodophenyl)acetic acid ≥98%
(3-Iodophenyl)acetic acid

Purity: 98% CAS No.: 1878-69-9 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Melting Point: 128-132° Beilstein Reference: 2501867

Formula: (C₆H₄I)CH₂COOH
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00046548
CAS Number: 1878-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR