2-Formyl-3-thiopheneboronic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

L15195.03 L15195.06
L15195.03EA 71.2 EUR
L15195.03 L15195.06
2-Formyl-3-thiopheneboronic acid ≥97%
2-Formyl-3-thiopheneboronic acid

Purity: 97% CAS No.: 4347-31-3 Molecular Formula: C5H5BO3S Formula Weight: 155.97 Storage Temperature: 2°C to 8°C Melting Point: ca 200° dec. Beilstein Reference: 4393584 MDL No.: MFCD01075678

Formula: C₅H₅BO₃S
Melting Pt: ∼ 200 °C (dec.)
MDL Number: MFCD01075678
CAS Number: 4347-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR