2-Chloro-6-nitrobenzamide ≥98%

Supplier: Thermo Fisher Scientific
Warning

L16949.06
L16949.06EA 0 EUR
L16949.06
2-Chloro-6-nitrobenzamide ≥98%
2-Chloro-6-nitrobenzamide

Purity: 98% CAS No.: 107485-64-3 Molecular Formula: C7H5ClN2O3 Formula Weight: 200.58 Melting Point: ca 185° Beilstein Reference: 3531056

Formula: C₇H₅ClN₂O₃
Melting Pt: ∼ 185 °C
Storage Temperature: Ambient
MDL Number: MFCD02093931
CAS Number: 107485-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR