Silicon dioxide 99.9%, powder 1,5 µm

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

L16987.14 L16987.22
L16987.14EA 88.4 EUR
L16987.14 L16987.22
Silicon dioxide 99.9%, powder 1,5 µm
Silicium dioxide

Purity: 99.9% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Odour: Odourless Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Density: 2,0
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR