1-Chloroisoquinoline 95%

Supplier: Thermo Fisher Scientific
Warning

L17831.03 L17831.06
L17831.03EA 38.7 EUR
L17831.03 L17831.06
1-Chloroisoquinoline 95%
1-Chloroisoquinoline

Purity: 95% CAS No.: 19493-44-8 Molecular Formula: C9H6ClN Formula Weight: 163.61 Flash Point: 110°(230°F) Melting Point: 33-35° Boiling Point: 105-108°/4mm Beilstein Reference: 2996 MDL No.: MFCD00024134

Formula: C₉H₆ClN
Boiling Pt: 105…108 °C (4 mmHg)
Melting Pt: 33…35 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00024134
CAS Number: 19493-44-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR