3-Formyl-4-methoxyphenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

L17850.03 L17850.06
L17850.03EA 29.9 EUR
L17850.03 L17850.06
3-Formyl-4-methoxyphenylboronic acid ≥98%
3-Formyl-4-methoxyphenylboronic acid

Purity: 98% CAS No.: 121124-97-8 Molecular Formula: C8H9BO4 Formula Weight: 179.97 Melting Point: ca 240° dec. Beilstein Reference: 7423998

Formula: C₈H₉BO₄
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02093661
CAS Number: 121124-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR