1,6-Dimethoxynaphthalene ≥98%

Supplier: Thermo Fisher Scientific

L18129.06 L18129.03
L18129.06EA 84.9 EUR
L18129.06 L18129.03
1,6-Dimethoxynaphthalene ≥98%
1,6-Dimethoxynaphthalene

Purity: 98+% CAS No.: 3900-49-0 Molecular Formula: C12H12O2 Formula Weight: 188.23 Melting Point: 58-59° Boiling Point: 123°/0.8mm MDL No.: MFCD00086330

Formula: C₁₂H₁₂O₂
Boiling Pt: 123 °C (0,8 mmHg)
Melting Pt: 58…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00086330
CAS Number: 3900-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR