3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%

Supplier: Thermo Fisher Scientific
Danger

L18180.06 L18180.03
L18180.06EA 356 EUR
L18180.06 L18180.03
3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
3,5-Bis(trifluoromethyl)-o-phenylenediamine

Purity: 97% CAS No.: 367-65-7 Molecular Formula: C8H6F6N2 Formula Weight: 244.14 Melting Point: 44-49° Beilstein Reference: 3344332 UN No.: 2811 MDL No.: MFCD01631430

Formula: C₈H₆F₆N₂
Melting Pt: 44…49 °C
Storage Temperature: Ambient
MDL Number: MFCD01631430
CAS Number: 367-65-7
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR