3,5-Bis(trifluoromethyl)pyridine ≥97%

Supplier: Thermo Fisher Scientific
Danger

L19447.03 L19447.MD
L19447.03EA 175 EUR
L19447.03 L19447.MD
3,5-Bis(trifluoromethyl)pyridine ≥97%
3,5-Bis(trifluoromethyl)pyridine

Purity: 97% CAS No.: 20857-47-0 Molecular Formula: C7H3F6N Formula Weight: 215.10 Flash Point: 100°(212°F) Melting Point: 35-38° Boiling Point: 117-118° Beilstein Reference: 1684576 MDL No.: MFCD00128904

Formula: C₇H₃F₆N
Boiling Pt: 117…118 °C
Melting Pt: 35…38 °C
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00128904
CAS Number: 20857-47-0
UN: 2810
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR