cis-Stilbeneboronic acid pinacol ester ≥99%

Supplier: Thermo Fisher Scientific

L19576.03
L19576.03EA 0 EUR
L19576.03
cis-Stilbeneboronic acid pinacol ester ≥99%
cis-Stilbeneboronic acid pinacol ester

Purity: 99% CAS No.: 264144-59-4 Molecular Formula: C20H23BO2 Formula Weight: 306.22 Melting Point: 92-94° Beilstein Reference: 9269561 MDL No.: MFCD03788734

Formula: C₂₀H₂₃BO₂
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD03788734
CAS Number: 264144-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR