2-Amino-3-formylpyridine 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Amino-3-pyridinecarboxaldehyde, 2-Aminonicotinaldehyde, 2-Aminopyridine-3-carboxaldehyde

L19620.03 L19620.06
L19620.03EA 73.1 EUR
L19620.03 L19620.06
2-Amino-3-formylpyridine 98%
2-Amino-3-formylpyridine

Purity: 98% CAS No.: 7521-41-7 Molecular Formula: C6H6N2O Formula Weight: 122.13 Storage Temperature: 2°C to 8°C Melting Point: 99-102° Beilstein Reference: 109598 MDL No.: MFCD01830382

Formula: C₆H₆N₂O
Melting Pt: 99…102 °C
MDL Number: MFCD01830382
CAS Number: 7521-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR