N-Boc-1,6-diaminohexane 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-tert-Butoxycarbonyl-1,6-hexanediamine, N-Boc-1,6-Diaminohexane

L19948.03 L19948.06 L19948.14
L19948.03EA 55 EUR
L19948.03 L19948.06 L19948.14
N-Boc-1,6-diaminohexane 98%
N-Boc-1,6-diaminohexane

Purity: 98% CAS No.: 51857-17-1 Molecular Formula: (CH3)3COCONH(CH2)6NH2 Formula Weight: 216.33 Flash Point: 110°(230°F) Refractive Index: 1.4620 Boiling Point: 106-110°/0.3mm Beilstein Reference: 2089264 UN No.: UN2735 MDL No.: MFCD00671489

Formula: C₁₁H₂₄N₂O₂
MW: 216.33 g/mol
Boiling Pt: 106…110 °C (0.3 mmHg)
Density: 0.965
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671489
CAS Number: 51857-17-1
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR