2-Nitrophenylboronic acid pinacol ester ≥98%

Supplier: Thermo Fisher Scientific
Warning

L19963.03 L19963.06
L19963.03EA 76.5 EUR
L19963.03 L19963.06
2-Nitrophenylboronic acid pinacol ester ≥98%
2-Nitrophenylboronic acid pinacol ester

Purity: 98+% CAS No.: 190788-59-1 Molecular Formula: C12H16BNO4 Formula Weight: 249.08 Melting Point: 42-44° Beilstein Reference: 7708824

Formula: C₁₂H₁₆BNO₄
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD02179447
CAS Number: 190788-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR