2-Bromo-5-iodopyridine 97

Supplier: Thermo Fisher Scientific
Warning

L20016.03 L20016.06
L20016.03EA 30.1 EUR
L20016.03 L20016.06
2-Bromo-5-iodopyridine 97
2-Bromo-5-iodopyridine

Purity: 97% CAS No.: 73290-22-9 Molecular Formula: C5H3BrIN Formula Weight: 283.89 Melting Point: 123-124° Beilstein Reference: 109100

Formula: C₅H₃BrIN
MW: 283.89 g/mol
Melting Pt: 123…124 °C
Storage Temperature: Ambient
MDL Number: MFCD03095201
CAS Number: 73290-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR