2-Bromo-3-nitropyridine ≥98%

Supplier: Thermo Fisher Scientific
Warning

L20019.03 L20019.06
L20019.03EA 31.4 EUR
L20019.03 L20019.06
2-Bromo-3-nitropyridine ≥98%
2-Bromo-3-nitropyridine

Purity: 98% CAS No.: 19755-53-4 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Melting Point: 122-125° Beilstein Reference: 124504

Formula: C₅H₃BrN₂O₂
Melting Pt: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00955613
CAS Number: 19755-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR