1-Bromo-2-butyne ≥98%

Supplier: Thermo Fisher Scientific
Warning

L20134.06 L20134.14
L20134.06EA 178 EUR
L20134.06 L20134.14
1-Bromo-2-butyne ≥98%
1-Bromo-2-butyne

Purity: 98% CAS No.: 3355-28-0 Molecular Formula: CH3C≡CCH2Br Formula Weight: 132.99 Flash Point: 36°(96°F) Refractive Index: 1.5080 Boiling Point: 123-125° Beilstein Reference: 605306 UN No.: 1993 MDL No.: MFCD00190233

Formula: C₄H₅Br
Boiling Pt: 123…125 °C
Density: 1,519
Flash Pt: 36 °C (96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190233
CAS Number: 3355-28-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR