Super drum, omologated
Supplier: LAMAPLAST
3209
LAMA3209EA 1500 EUR
LAMA3209
Super drum, omologated
Barrels, Kegs, Drums
These hermetic safety container are produced by rotational system in high density polyethylene with an high molecular weight. Suitable for the transport and storing of metal drums containing harmful, toxic substances.
  • Resistant to impact, chemical and atmospheric agents
  • Double wall lid with dutral seal and stainless steel iron ring for a perfect hermetic seal
Order Now
SPECIFICATIONS
Environmentally Preferable