Cooling circulation thermostats, PRO
Supplier: LAUDA
L000021 L000022 L000023 L000024 L000102 L000104 L000122 L000124 L000182 L000184 L000202 L000204 L002608 L002609 L002610 L002611
461-0953EA 9540 EUR
461-0953 461-0954 461-0955 461-0956 LAUDL000102 LAUDL000104 LAUDL000122 LAUDL000124 LAUDL000182 LAUDL000184 LAUDL000202 LAUDL000204 LAUDL002608 LAUDL002609 LAUDL002610 LAUDL002611
Cooling circulation thermostats, PRO
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The PRO cooling circulation thermostats are ideal for external applications where rapid temperature changes are required. Their cooling output of 0,6 to 1,5 kW, combined with a very small filling volume enable these rapid temperature changes.
  • Touchscreen can be operated with gloves
  • Low filling volume combined with Varioflex pump ensures rapid temperature changes
  • Programmer with one program, 20 segments whereas 100 programs, with 50 segments for Command unit
  • Enables ramp function for easy computation
  • Hybrid cooling for cooling the refrigeration machine with
More Product Information