Process Thermostats, Variocool
Supplier: LAUDA
L000983 L000985 L000664 L000665 L000677 L000678 L000728 L000729 L000730 L000746 L000747 L000748 L000949 L000951 L000953
LAUDL000983EA 31650 EUR
LAUDL000983 LAUDL000985 LAUDL000664 LAUDL000665 LAUDL000677 LAUDL000678 LAUDL000728 LAUDL000729 LAUDL000730 LAUDL000746 LAUDL000747 LAUDL000748 LAUDL000949 LAUDL000951 LAUDL000953
Process Thermostats, Variocool
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The compact Variocool circulating chillers are ideal for demanding temperature control tasks in laboratories, mini plants and production. The units have a colour TFT screen and membrane keyboard, a USB interface and an integrated alarm which are easily accessable on the front of the unit. They also feature an electronic fill gauge on the display and low fluid level alarm. Models can be either air or water cooled, all have lockable castors. A wide range of factory fitted options and interfaces which can be retrofitted give the user greater flexibility and control. Options include high powerMore Product Information