Freeze dryer, console, FreeZone®
Supplier: LABCONCO
700611050 700611150 700611250 700611350 700612050 700612150 700612250 700612350 700621050 700621150 700622050 700622150 701211050 701211150 701211250 701212050 701212150 701212250 701221050 701221150 701222050 701222150 701811050 701811150 701811250 701812050 701812150 701812250 701821050 701821150 701822050 701822150 710611050 710611150 710611250 710612050 710612150 710612250 710621050 710621150 710622050 710622150 711211050 711211150 711211250 711212050 711212150 711212250 711221050 711221150 711222050 711222150
LBCP700611050EA 0 EUR
LBCP700611050 LBCP700611150 LBCP700611250 LBCP700611350 LBCP700612050 LBCP700612150 LBCP700612250 LBCP700612350 LBCP700621050 LBCP700621150 LBCP700622050 LBCP700622150 LBCP701211050 LBCP701211150 LBCP701211250 LBCP701212050 LBCP701212150 LBCP701212250 LBCP701221050 LBCP701221150 LBCP701222050 LBCP701222150 LBCP701811050 LBCP701811150 LBCP701811250 LBCP701812050 LBCP701812150 LBCP701812250 LBCP701821050 LBCP701821150 LBCP701822050 LBCP701822150 LBCP710611050 LBCP710611150 LBCP710611250 LBCP710612050 LBCP710612150 LBCP710612250 LBCP710621050 LBCP710621150 LBCP710622050 LBCP710622150 LBCP711211050 LBCP711211150 LBCP711211250 LBCP711212050 LBCP711212150 LBCP711212250 LBCP711221050 LBCP711221150 LBCP711222050 LBCP711222150
Freeze dryer, console, FreeZone®
Freeze Dryers
FreeZone® console freeze dryers with or without stoppering tray dryers are designed for lyophilising moderate to large loads and stoppering vials or serum bottles under vacuum. The console features a front viewing window and interior space for a vacuum and accommodation for a full size tray dryer such as the FreeZone® stoppering tray dryer pump (sold separately).
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ end
More Product Information