Water purification system, Water Pro RO
Supplier: LABCONCO
9075030
LBCP9075030EA 0 EUR
LBCP9075030
Water purification system, Water Pro RO
Water Purification Systems Pure and Ultra Pure Water Systems
WaterPro RO stations feature large capacity filters and membrane to deliver high-quality reverse osmosis (RO) purified water (type III). RO purified water is ideal for laboratory applications such as reagent preparation and glassware rinsing. The units be connected to glassware washers to provide purified rinse water.
  • Timed dispense from 1 to 99,9 minutes for unattended operation
  • Attractive and durable Type 304 stainless steel front panel; epoxy-coated steel cabinet
  • Large capacity white polypropylene bowls with threaded heads
  • Integral 17 liter PE storage
More Product Information