Disc Dispensers
Supplier: LIOFILCHEM
91203 91200
LIOF91203EA 1160 EUR
LIOF91203 LIOF91200
Disc Dispensers
Petri Dish Fillers
Disc dispenser is a device that allows to place antibiotic discs on the agarsurface of petri dishes.
  • Airtight container with desiccant
  • Label for the identification of antibiotics positioned on top of the container
  • Block-cartridges system positioned at the base of the plunger
  • Height adjustable base (1 cm)
The discs are positioned at equidistance along a circumference of about 16 cm. Each disk is pressed on the agar surface with a pressure sufficient to ensure a homogeneous diffusion of the antibiotic.
Disc dispenser in its container with desiccantMore Product Information