Water, Ecotainer® endotoxin-free, sterile

Supplier: Local

Synonyms: Hydrogen oxide

0082479E
LOCA0082479EEA 0 EUR
LOCA0082479E
Water, Ecotainer® endotoxin-free, sterile
Water

Rinsing solution in PP bottle.


  • Sterile
  • Pyrogen free

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR