Monoparameter photometers, Lovibond® MD 100

Supplier: Lovibond

276010 276000 276030 276035 276110 276160 276040 276005 276050 276060 276080 276090 276100 276170 276190 276200 276116 276020 276025 276120 276210 276205
705-0964EA 553 EUR
705-0964 LOVI276000 LOVI276030 LOVI276035 LOVI276110 LOVI276160 LOVI276040 LOVI276005 LOVI276050 LOVI276060 LOVI276080 LOVI276090 LOVI276100 LOVI276170 LOVI276190 LOVI276200 LOVI276116 LOVI276020 LOVI276025 LOVI276120 LOVI276210 LOVI276205
Monoparameter photometers, Lovibond® MD 100
Environmental Test Kits

The MD 100 range of photometers are portable handheld instruments with a simple button layout, robust waterproof design and large backlit display that are ideal for testing in the field or laboratory conditions.


  • Accurate (3% FS), reproducible results using high quality interference filters with long-life LEDs
  • Internal ring memory for 16 data sets
  • Real time clock and date
  • Optional infrared interface for transfer of test data to PC or printer


Delivery information: Instrument supplied with carry case, 4× AAA batteries, 3× round glass vials with lids, relevant reagents, and instruction manual.

Order Now

SPECIFICATIONS

Model MD 100
Environmentally Preferable
Accuracy 3% FS (T=20...25°C)
Wavelengths 430, 530, 560, 580, 610, 660 nm
Wavelength accuracy ±1 nm
Automatic switch-off
Battery life 17 h or 5000 tests
Display LCD, backlit
Interfaces Infrared interface for test data transfer
Keypad 4 button membrane keypad
Operating humidity 30 - 90% RH
Operating temperature 5…40 °C
Optics LED, interference filters (IF) and photo sensor in transparent sample chamber
Power supply 4× AAA batteries
Storage capacity 16 data sets
W×D×H 155×75×35 mm
Weight ~0,26 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR