Iodoacetamide ≥98%, Proteomics Grade

Supplier: VWR Chemicals
Warning

Synonyms: 2-Iodoacetamide

M216-30GEA 276 EUR
M216-30G
Iodoacetamide ≥98%, Proteomics Grade
Iodoacetamide
Formula: ICH₂CONH₂
MW: 184.96 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 91…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2928
ADR: 6.1 II

Order Now

Specification Test Results

Loss on drying <1%
Melting point 90 − 93 °C
Protease none detected
Purity >98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR