Lucigenin chemiluminescence reagent, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ100

M8010-1G M8010-500MG M8010-5G
M8010-1GEA 469 EUR
M8010-1G M8010-500MG M8010-5G
Lucigenin chemiluminescence reagent, Sigma-Aldrich®
Lucigenin

Reagent used for the generation of chemiluminescence in the presence of a reductant and oxygen; it is employed in numerous chemiluminescent assays. Lucigenin is specific for superoxide anion radical.

Formula: C₂₈H₂₂N₄O₆
Melting Pt: 245,..250 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00011925
CAS Number: 2315-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f7dd389c39e0a67bb32902b5b7b973752436476&vid=b97cf0a1dc049e217bc8b834a739256b4cd344b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour)
Yellow with an Orange to Brown Cast
Conforms
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
TLC Consistent with Past Lots
Solubility (Turbidity)
C = 40 mg/4 ml, Acetic Acid
Clear to Hazy
Solubility (Colour)
Yellow to Very Dark Yellow
Conforms to Requirements


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR