Crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
70208.36 70217.36 70204.36 70205.36 70206.36 70209.1 70210.36 70215.36 70216.36 702261 70254 702917 702918 702924 702263 702263.2 702020 702086 702540
MANA70208.36EA 124 EUR
MANA70208.36 391-0101 70204. MANA70205.36 MANA70206.36 MANA70209.1 MANA70210.36 70215.36 70216.36 MANA702261 MANA70254 MANA702917 MANA702918 MANA702924 MANA702263 MANA702263.2 MANA702020 MANA702086 MANA702540
Crimp neck vials, N 20
Vials
These crimp neck vials with different capacities are suitable for a large number of autosamplers included headspace GC applications.
  • Made of clear and amber glass for light-sensitive substances
  • Flat and rounded bottom
  • Beveled HS and flat DIN crimp necks available
  • Label for convenient handling
  • Graduation for defined filling
Useable for the common GC headspace autosamplers of Agilent, CTC, PerkinElmer (Combi PAL, TurboMatrix HS 16, 40, 110), Shimadzu (AOC-5000, AOC-6000, HS-20, HS-20Trap, HT200H), Thermo Scientific (Combi PAL), Varian (Agilent - CombiMore Product Information