Bulk media for HPLC, POLYGOSIL®
Supplier: MACHEREY-NAGEL
711010.10 711010.100 711020.10 711020.100 711280.10 711280.100 711300.10 711300.100 711310.10 711310.100 711320.10 711320.100 711330.10 711330.100 711340.10 711340.100 711350.10 711350.100 711360.10 711360.100 711370.10 711370.100 711380.10 711380.100 711390.10 711390.100 711420.10 711420.100 711430.10 711430.100 711510.10 711510.100 711520.10 711520.100 711530.10 711530.100 711560.10 711560.100 711570.10 711570.100 711580.10 711580.100 711600.10 711600.100 711680.10 711680.100 711710.10 711710.100 711890.10 711890.100 711991.10 711991.100 711992.10 711992.100
MANA711010.10EA 67.2 EUR
MANA711010.10 MANA711010.100 MANA711020.10 MANA711020.100 MANA711280.10 MANA711280.100 MANA711300.10 MANA711300.100 MANA711310.10 MANA711310.100 MANA711320.10 MANA711320.100 MANA711330.10 MANA711330.100 MANA711340.10 MANA711340.100 MANA711350.10 MANA711350.100 711360.10 MANA711360.100 MANA711370.10 MANA711370.100 MANA711380.10 MANA711380.100 MANA711390.10 MANA711390.100 MANA711420.10 MANA711420.100 MANA711430.10 MANA711430.100 MANA711510.10 MANA711510.100 MANA711520.10 MANA711520.100 MANA711530.10 MANA711530.100 MANA711560.10 MANA711560.100 MANA711570.10 MANA711570.100 MANA711580.10 MANA711580.100 MANA711600.10 MANA711600.100 MANA711680.10 MANA711680.100 MANA711710.10 MANA711710.100 MANA711890.10 MANA711890.100 MANA711991.10 MANA711991.100 MANA711992.10 MANA711992.100
Bulk media for HPLC, POLYGOSIL®
Chromatography Sorbents HPLC Sorbents
Irregular silica phase for analytical HPLC.
  • Range of pore sizes 60, 100, 300 and 1000 Å
  • Available particle sizes 5, 7 and 10 µm
  • Different modifications
  • pH stability 2 to 8
The irregular silica POLYGOSIL® can be used for different analytical applications.
Order Now
SPECIFICATIONS
Environmentally Preferable