Flash chromatography cartridges, CHROMABOND® Flash RS
Supplier: MACHEREY-NAGEL
732460 732461 732462 732800 732803 732804 732805 732810 732811 732812 732809 732808 732806 732807 732802 732801 732762 732767 732718 732716 732717 732719 732720 732760 732761 732763 732764 732765 732766 732813 732814 732815 732816 732817 732818 732819 732850 732463 732464 732465 732466 732467 732700 732701 732702 732703 732704 732705 732706 732707 732708 732711 732712 732713 732714 732715 732709
MANA732460EA 292 EUR
MANA732460 MANA732461 MANA732462 MANA732800 MANA732803 MANA732804 554-6564 554-6569 554-6570 554-6571 554-6568 554-6567 554-6565 554-6566 554-6563 554-6562 554-6556 554-6561 554-6552 554-6550 554-6551 554-6553 MANA732720 554-6554 554-6555 554-6557 554-6558 554-6559 554-6560 554-6572 554-6573 554-6574 554-6575 554-6576 554-6577 554-6578 554-6579 MANA732463 MANA732464 MANA732465 MANA732466 MANA732467 554-6535 554-6536 554-6537 554-6538 554-6539 554-6540 554-6541 554-6542 554-6543 554-6545 554-6546 554-6547 554-6548 554-6549 554-6544
Flash chromatography cartridges, CHROMABOND® Flash RS
Chromatography Columns
These ready-to-use Flash cartridges are designed for use in the Teledyne Isco® Companion® and other Teledyne Isco® CombiFlash® systems without additional connectors or capillaries or as stand-alone version for any pump/detector combinations.
Safe hardware
- Heavy-duty polypropylene cartridges
- High pressure stability up to 21 bar (RS 4 to RS 40), 15 bar (RS 80 to RS 120), 12 bar (RS 200 to RS 1600) and 7 bar (RS 3000)
- Female Luer lock inlet / male Luer outlet (RS 4 to RS 330) and Maxi Luers (RS 800 to RS 3000)
Considerable program of efficient phases
- Irregular unmodified silica SiOH (More Product Information