Cellulose MN 100 adsorbents for column chromatography

Supplier: MACHEREY-NAGEL

815050.1 815050.25 815050.5
MANA815050.1EA 41.6 EUR
MANA815050.1 MANA815050.25 MANA815050.5
Cellulose MN 100 adsorbents for column chromatography
Cellulose

Native fibrous cellulose with an average degree of polymerisation 620 to 680.


  • For separation of hydrophilic substances
  • Fiber length (85%): 20 to 100 μm
  • Specific surface according to Blaine ~6500 cm²/g

Formula: (C₁₂H₂₂O₁₁)n
Melting Pt: 260…270 °C
Density: 1,27…1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Order Now

Specification Test Results

Residue on Ignition <10000 ppm
Fe <20 ppm
Cu <5 ppm
P <7 ppm
Dichloromethane Extract <0.20%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR