Counting chambers, Howard
Supplier: MARIENFELD
0610940 0611040
MARI0610940EA 239 EUR
MARI0610940 MARI0611040
Counting chambers, Howard
Slides Counting Chambers
Counting chambers designed for the investigation of fruit juices.
Ø ring = 18 - 22 mm
Order Now
SPECIFICATIONS
Environmentally Preferable