Diamond pen with wooden handle
Supplier: MARIENFELD
6120300
MARI6120300EA 40.1 EUR
MARI6120300
Diamond pen with wooden handle
Pens Ink Pens
For permanent labelling of microscope slides and other glass products. Indelibly engraved markings comply particularly with the guidelines of GLP.
  • Resistant to humidity, heat, cold and solvents
  • With wooden handle