Brilliant cresyl blue solution_SC stain_biological, Microscopy, Sigma-Aldrich®

Supplier: Merck

Synonyms: CI 51010, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1), Brilliant cresyl blue zinc chloride double salt, Brilliant Blue C, BCB

1.01384.0100 1.01384.9025
1.01384.0100EA 52.4 EUR
1.01384.0100 MERC1.01384.9025
Brilliant cresyl blue solution_SC stain_biological, Microscopy, Sigma-Aldrich®
Brilliant Cresyl Blue

The Brilliant cresyl blue solution is used in hematology as reagent for the staining of blood for the subsequent manual counting of blood cells. The regenerative capacity of erythrocytes can be monitored with the ratio of reticulocytes. In addition, the determination of the Substantia granulo-filamentosa (ribonucleoproteins) is possible with fresh, unfixed young erythrocytes (in a supra-vitral staining).


The Brilliant cresyl blue solution is an IVD & CE product and 100 ml of the solution are capable of staining up to 5000 slides.

Formula: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
Density: 1.01 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148901
CAS Number: 81029-05-2
IVDLogo

Order Now

Specification Test Results

Application test (Reticulocyte staining) passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR