EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) for synthesis

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.00907.0001 8.00907.0005 8.00907.0025
8.00907.0001EA 22.4 EUR
8.00907.0001 MERC8.00907.0005 MERC8.00907.0025
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) for synthesis
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Formula: C₈H₁₇N₃·HCl
Melting Pt: 112…113 °C
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR