Grounding cables, Super Erecta®
Supplier: Metro International
ASK16S GCB16SA
METOASK16SEA 31.7 EUR
METOASK16S METOGCB16SA
Grounding cables, Super Erecta®
Grounders Grounding Wires
These grounding cables provide an effective method of electrostatic discharge for any Metro Super Erecta unit (stationary or mobile) when used in conjunction with aluminum split sleeves or ESD kit and conductive casters.
  • Cables ensure a proper path to ground when used with ESD flooring
  • Grounding cable with universal bracket fits wire and solid shelves, as well as open-wire frames