Ion selective electrodes and half-cells
Supplier: Mettler - Toledo
51340263 51107672 51107673 51107674 51107675 51107676 51107678 51107680 51107681 51107686 51107687 51107688 51107689 51107690 51107692 51107694 51107695 51107870 51107871 51107873 51107874 51089935
662-2803EA 549 EUR
662-2803 METT51107672 METT51107673 METT51107674 METT51107675 METT51107676 METT51107678 METT51107680 METT51107681 METT51107686 METT51107687 METT51107688 METT51107689 METT51107690 METT51107692 METT51107694 METT51107695 METT51107870 METT51107871 METT51107873 METT51107874 METT51089935
Ion selective electrodes and half-cells
Electrodes ISE Electrodes
These ion-selective half-cells (exception: Sodium-sensitive electrode) consist of two elements: a universal shaft and an ion-specific membrane module.
  • Available as half-cells (require external reference electrode) or as electrodes with built-in reference
  • Each ion-selective electrode (ISE) and each membrane module is tested wet-chemically
  • Each ISE is issued with its own serial number and quality certificate
  • Electrodes are filled with electrolyte and ready for use
  • Membrane kit comes with an identification ring (ID ring) and an adapter sleeve which will fit
More Product Information