Pressure Relief Valves, 316 Stainless Steel
Supplier: Avantor Fluid Handling
03245-40
MFLX03245-40EA 868 EUR
MFLX03245-40
Pressure Relief Valves, 316 Stainless Steel
Valves Pressure-Relief Valves
Easily control your outlet pressure.
  • Constructed of durable 316 stainless steel (SS)
Use for liquid applications with flow rates up to 15 GPM. For pressures from 40 to 150 psig. Valves have 1/2" NPT(F); orifice size is 5/16".